missing translation for 'onlineSavingsMsg'
Learn More

(6-bromopyrid-2-yl)tributylstannane, 95%, Thermo Scientific™

Catalog Number p-3880832
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
189083-81-6
C17H30BrNSn
447.048
MFCD11706981
ZRPIKMNUVOBWBE-UHFFFAOYSA-N
2-bromo-6-tributylstannyl pyridine, pyridine, 2-bromo-6-tributylstannyl, 6-bromopyrid-2-yl tributylstannane, acmc-20ao22, 2-bromopyridin-6-yl tributylstannane, 2-bromo-6-tributylstannyl-pyridine, 2-bromo-6-tributylstannyl pyridine h-nmr
24787029
(6-bromopyridin-2-yl)-tributylstannane
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)Br
This item is not returnable. View return policy
189083-81-6
C17H30BrNSn
447.048
MFCD11706981
ZRPIKMNUVOBWBE-UHFFFAOYSA-N
2-bromo-6-tributylstannyl pyridine, pyridine, 2-bromo-6-tributylstannyl, 6-bromopyrid-2-yl tributylstannane, acmc-20ao22, 2-bromopyridin-6-yl tributylstannane, 2-bromo-6-tributylstannyl-pyridine, 2-bromo-6-tributylstannyl pyridine h-nmr
24787029
(6-bromopyridin-2-yl)-tributylstannane
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)Br
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 447.05
Percent Purity 95%
Chemical Name or Material (6-Bromopyrid-2-yl)tributylstannane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'