missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopicolinic acid, 98%, Acros Organics™

Brand:  Acros Organics 379930250

Code : 29

Additional Details : CAS Number : 21190-87-4 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11976331

  • £379.90 / 25g


Estimated Shipment
18-05-2018
Add to basket

Specifications

Chemical Identifiers

6-Bromopicolinic acid
98%
202.01
98%
C6H4BrNO2

Specifications

6-Bromopicolinic acid
Authentic
192.0° to 194.0°C
Powder
Off-white to tan
C6H4BrNO2
21190-87-4
Glass bottle
25g
98%
98%
202.01
Videos
SDS