missing translation for 'onlineSavingsMsg'
Learn More

6-Bromonicotinic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-4228463
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6311-35-9
C6H4BrNO2
202.01
MFCD01927100
JDJBRMNTXORYEN-UHFFFAOYSA-N
6-bromonicotinic acid, 6-bromo nicotinic acid, 2-bromo-5-pyridinecarboxylic acid, 2-bromopyridine-5-carboxylic acid, 6-bromo-3-pyridine carboxylic acid, 6-bromonicotinicacid, 6-bromo-nicotinic acid, 6-bromo-3-pyridinecarboxylic acid, 2-bromo-5-pyridine carboxylic acid, 3-pyridinecarboxylic acid, 6-bromo
238932
6-bromopyridine-3-carboxylic acid
OC(=O)C1=CC=C(Br)N=C1
This item is not returnable. View return policy
6311-35-9
C6H4BrNO2
202.01
MFCD01927100
JDJBRMNTXORYEN-UHFFFAOYSA-N
6-bromonicotinic acid, 6-bromo nicotinic acid, 2-bromo-5-pyridinecarboxylic acid, 2-bromopyridine-5-carboxylic acid, 6-bromo-3-pyridine carboxylic acid, 6-bromonicotinicacid, 6-bromo-nicotinic acid, 6-bromo-3-pyridinecarboxylic acid, 2-bromo-5-pyridine carboxylic acid, 3-pyridinecarboxylic acid, 6-bromo
238932
6-bromopyridine-3-carboxylic acid
OC(=O)C1=CC=C(Br)N=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 179°C to 181°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 202.01
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromonicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 6-Bromonicotinic Acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'