Learn More
Thermo Scientific Maybridge 6-(Bromomethyl)-2-methylquinoline, 1g, 97%
Brand: Thermo Scientific Maybridge CC28408DA
6-(Bromomethyl)-2-methylquinoline, 1g, 97%
Code : 31
Additional Details : CAS Number : 141848-60-4 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC28408DA
6-(Bromomethyl)-2-methylquinoline, 1g, 97%
Code : 31
Additional Details : CAS Number : 141848-60-4 Weight : 0.05100kg