missing translation for 'onlineSavingsMsg'
Learn More

6-Bromohexanoyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number 11314649
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
22809-37-6
C6H10BrClO
213.499
MFCD00000761
HBPVGJGBRWIVSX-UHFFFAOYSA-N
hexanoyl chloride, 6-bromo, 6-bromohexanoic acid chloride, 6-bromohexanoylchloride, 6-bromocaproyl chloride, bromohexanoyl chloride, zlchem 435, acmc-209fyy, 6-bromo-hexanoylchloride, 6-bromo-hexanoyl chloride, 6-bromanylhexanoyl chloride
89848
6-bromohexanoyl chloride
C(CCC(=O)Cl)CCBr
This item is not returnable. View return policy
22809-37-6
C6H10BrClO
213.499
MFCD00000761
HBPVGJGBRWIVSX-UHFFFAOYSA-N
hexanoyl chloride, 6-bromo, 6-bromohexanoic acid chloride, 6-bromohexanoylchloride, 6-bromocaproyl chloride, bromohexanoyl chloride, zlchem 435, acmc-209fyy, 6-bromo-hexanoylchloride, 6-bromo-hexanoyl chloride, 6-bromanylhexanoyl chloride
89848
6-bromohexanoyl chloride
C(CCC(=O)Cl)CCBr

6-Bromohexanoyl chloride is used as intermediates, organic synthesis, chemical research, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromohexanoyl chloride is used as intermediates, organic synthesis, chemical research, agrochemicals and dyestuff.

Solubility
Reacts with water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed. Moisture Sensitive
TRUSTED_SUSTAINABILITY
Density 1.397
Boiling Point 119°C to 120°C (9 mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Linear Formula Br(CH2)5COCl
Refractive Index 1.4875
Quantity 5 g
UN Number UN3265
Beilstein 1749739
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 213.5
Percent Purity 97%
Chemical Name or Material 6-Bromohexanoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'