missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromohexanoic acid, 98%, Thermo Scientific Chemicals
27.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 32.0°C to 35.0°C |
Color | Orange to White |
Boiling Point | 165.0°C to 170.0°C (20.0 mmHg) |
Flash Point | 67°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | Br(CH2)5CO2H |
Quantity | 25 g |
Beilstein | 02,II,287 |
Show More |
Safety and Handling
Product Identifier
- 6-Bromohexanoic acid
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'