missing translation for 'onlineSavingsMsg'
Learn More

6-Bromochroman, 97%, Thermo Scientific™

Catalog Number p-3880702
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
3875-78-3
C9H9BrO
213.074
MFCD10698725
KPFDABVKWKOIME-UHFFFAOYSA-N
6-bromochroman, 6-bromo-chroman, 6-bromochromane, 6-bromo-3,4-dihydro-2h-1-benzopyran, 2h-1-benzopyran,6-bromo-3,4-dihydro, 6-bromanyl-3,4-dihydro-2h-chromene, 2h-1-benzopyran, 6-bromo-3,4-dihydro
10856814
6-bromo-3,4-dihydro-2H-chromene
C1CC2=C(C=CC(=C2)Br)OC1
This item is not returnable. View return policy
3875-78-3
C9H9BrO
213.074
MFCD10698725
KPFDABVKWKOIME-UHFFFAOYSA-N
6-bromochroman, 6-bromo-chroman, 6-bromochromane, 6-bromo-3,4-dihydro-2h-1-benzopyran, 2h-1-benzopyran,6-bromo-3,4-dihydro, 6-bromanyl-3,4-dihydro-2h-chromene, 2h-1-benzopyran, 6-bromo-3,4-dihydro
10856814
6-bromo-3,4-dihydro-2H-chromene
C1CC2=C(C=CC(=C2)Br)OC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 213.07
Percent Purity 97%
Chemical Name or Material 6-Bromochroman
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'