missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-8-fluoroquinoline, 97%, Thermo Scientific™

Catalog Number 15289796
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
220513-46-2
C9H5BrFN
226.05
DDCOYWOYIUEOHQ-UHFFFAOYSA-N
acmc-1crnu, 6-bromo-8-fluoro-quinoline, 6-bromo-8-fluoroquinoline, 6-bromo-8-fluoro-1-azanaphthalene
46738885
6-bromo-8-fluoroquinoline
C1=CC2=CC(=CC(=C2N=C1)F)Br
This item is not returnable. View return policy
220513-46-2
C9H5BrFN
226.05
DDCOYWOYIUEOHQ-UHFFFAOYSA-N
acmc-1crnu, 6-bromo-8-fluoro-quinoline, 6-bromo-8-fluoroquinoline, 6-bromo-8-fluoro-1-azanaphthalene
46738885
6-bromo-8-fluoroquinoline
C1=CC2=CC(=CC(=C2N=C1)F)Br
Melting Point 103°C to 106°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 226.05
Percent Purity 97%
Chemical Name or Material 6-Bromo-8-fluoroquinoline
compliance-icons
Product Identifier
  • 6-Bromo-8-fluoroquinoline, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'