missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-pyridinecarboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog Number p-4813255
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
149806-06-4
C6H4BrNO
186.01
MFCD04115419
PVUKGNBRJFTFNJ-UHFFFAOYSA-N
6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
11769234
6-bromopyridine-3-carbaldehyde
BrC1=CC=C(C=O)C=N1
This item is not returnable. View return policy
149806-06-4
C6H4BrNO
186.01
MFCD04115419
PVUKGNBRJFTFNJ-UHFFFAOYSA-N
6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
11769234
6-bromopyridine-3-carbaldehyde
BrC1=CC=C(C=O)C=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 104.0°C to 110.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 186
Percent Purity 95%
Chemical Name or Material 6-Bromo-3-pyridinecarboxaldehyde
compliance-icons
Product Identifier
  • 6-Bromo-3-pyridinecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'