missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-pyridinecarboxaldehyde, 95%, Acros Organics

Brand:  Acros Organics 399850050

Code : 29

Additional Details : CAS Number : 149806-06-4 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 10460504

  • £135.10 / 5g


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

6-Bromo-3-pyridinecarboxaldehyde
95%
C6H4BrNO
186

Specifications

6-Bromo-3-pyridinecarboxaldehyde
Glass bottle
5g
95%
186
149806-06-4
104.0° to 110.0°C
95%
C6H4BrNO
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Bromo-3-pyridinecarboxaldehyde

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS