missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-butyrylcoumarin, 97%, Thermo Scientific™

Catalog Number p-7027933
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2199-83-9
C13H11BrO3
295.132
MFCD00052894
RDBKKAPCFCJFND-UHFFFAOYSA-N
6-bromo-3-butyryl-2h-chromen-2-one, maybridge1_001495, acmc-1cbq8, 6-bromo-3-butyrylcoumarin, 2h-1-benzopyran-2-one,6-bromo-3-1-oxobutyl
2799663
6-bromo-3-butanoylchromen-2-one
CCCC(=O)C1=CC2=C(C=CC(=C2)Br)OC1=O
This item is not returnable. View return policy
2199-83-9
C13H11BrO3
295.132
MFCD00052894
RDBKKAPCFCJFND-UHFFFAOYSA-N
6-bromo-3-butyryl-2h-chromen-2-one, maybridge1_001495, acmc-1cbq8, 6-bromo-3-butyrylcoumarin, 2h-1-benzopyran-2-one,6-bromo-3-1-oxobutyl
2799663
6-bromo-3-butanoylchromen-2-one
CCCC(=O)C1=CC2=C(C=CC(=C2)Br)OC1=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 163°C to 165°C
Quantity 1 g
Formula Weight 279.14
Percent Purity 97%
Chemical Name or Material 6-Bromo-3-butyrylcoumarin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'