missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Bromo-3-(4-oxycyclohexen-1-yl)indole, 95%

Brand:  Alfa Aesar™ H63679.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15430499

  • £478.06 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

6-Bromo-3-(4-oxycyclohexen-1-yl)indole
290.16
MFCD28126775
4-(6-Bromo-3-indolyl)cyclohex-3-en-1-one

Specifications

6-Bromo-3-(4-oxycyclohexen-1-yl)indole
5g
290.16
4-(6-Bromo-3-indolyl)cyclohex-3-en-1-one
Solid
95%
MFCD28126775
Videos
SDS