missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole, 97%

Brand:  Alfa Aesar™ H63660.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15488426

  • £59.78 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
334.21
MFCD28098698
6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole

Specifications

6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
97%
334.21
6-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
Solid
1g
MFCD28098698
Videos
SDS