missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-methyl-2H-indazole, 97%, Thermo Scientific Chemicals

Catalog Number p-4813253
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
590417-95-1
C8H7BrN2
211.06
BVYFYDANLZQCPV-UHFFFAOYSA-N
6-bromo-2-methyl-2h-indazole, 2-methyl-6-bromoindazole, 2h-indazole, 6-bromo-2-methyl, pubchem20671, acmc-209m9q, boron-mol bm788, 6-bromo-2-methyl-indazole, 2h-indazole,6-bromo-2-methyl
22558624
6-bromo-2-methylindazole
CN1C=C2C=CC(=CC2=N1)Br
This item is not returnable. View return policy
590417-95-1
C8H7BrN2
211.06
BVYFYDANLZQCPV-UHFFFAOYSA-N
6-bromo-2-methyl-2h-indazole, 2-methyl-6-bromoindazole, 2h-indazole, 6-bromo-2-methyl, pubchem20671, acmc-209m9q, boron-mol bm788, 6-bromo-2-methyl-indazole, 2h-indazole,6-bromo-2-methyl
22558624
6-bromo-2-methylindazole
CN1C=C2C=CC(=CC2=N1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 85°C to 87°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-methyl-2H-indazole
compliance-icons
Product Identifier
  • 6-Bromo-2-methyl-2H-indazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'