missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-methyl-1H-indole, ≥97%, Thermo Scientific™

Catalog Number p-3174841
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
125872-95-9
C9H8BrN
210.07
MFCD08543444
PXHJDPPKNUGKPM-UHFFFAOYSA-N
6-bromo-1-methyl-1h-indole, 1h-indole,6-bromo-1-methyl, acmc-20eb8h, 6-bromo-1-methyl indole, 1-methyl-6-bromo-1h-indole, 1h-indole, 6-bromo-1-methyl
15546854
CN1C=CC2=CC=C(Br)C=C12
This item is not returnable. View return policy
125872-95-9
C9H8BrN
210.07
MFCD08543444
PXHJDPPKNUGKPM-UHFFFAOYSA-N
6-bromo-1-methyl-1h-indole, 1h-indole,6-bromo-1-methyl, acmc-20eb8h, 6-bromo-1-methyl indole, 1-methyl-6-bromo-1h-indole, 1h-indole, 6-bromo-1-methyl
15546854
CN1C=CC2=CC=C(Br)C=C12
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 6-bromo-1-methylindole
Formula Weight 210.07
Percent Purity 97%
Chemical Name or Material 6-Bromo-1-methyl-1H-indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'