Learn More
6-Bromo-1-methyl-1H-indazole, 97%, ACROS Organics™
Brand: Acros Organics 444600050
Code : 29
Additional Details : CAS Number : 590417-94-0 Weight : 0.06000kg
Quantity | 5g |
---|
Chemical Identifiers
590417-94-0 | |
211.06 | |
6-bromo-1-methyl-1h-indazole, 1-methyl-6-bromoindazole, 1h-indazole, 6-bromo-1-methyl, pubchem19529, acmc-209m9p, 6-bromo-1-methyl-indazole, indazole, 6-bromo-1-methyl | |
6-bromo-1-methylindazole |
C8H7BrN2 | |
SVVSOIGNROPKMS-UHFFFAOYSA-N | |
22558675 | |
CN1C2=C(C=CC(=C2)Br)C=N1 |
Specifications
6-Bromo-1-methyl-1H-indazole | |
5g | |
96% min. (GC) | |
6-bromo-1-methyl-1h-indazole, 1-methyl-6-bromoindazole, 1h-indazole, 6-bromo-1-methyl, pubchem19529, acmc-209m9p, 6-bromo-1-methyl-indazole, indazole, 6-bromo-1-methyl | |
CN1C2=C(C=CC(=C2)Br)C=N1 | |
211.06 | |
211.06 |
48°C to 50°C | |
590417-94-0 | |
C8H7BrN2 | |
SVVSOIGNROPKMS-UHFFFAOYSA-N | |
6-bromo-1-methylindazole | |
22558675 | |
97% |
Safety and Handling
- 6-Bromo-1-methyl-1H-indazole
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.