missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-hexene, 97%, Thermo Scientific Chemicals

Catalog Number 10551504
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2695-47-8
C6H11Br
163.06
MFCD00000269
RIMXEJYJXDBLIE-UHFFFAOYSA-N
6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene
75906
6-bromohex-1-ene
C=CCCCCBr
This item is not returnable. View return policy
2695-47-8
C6H11Br
163.06
MFCD00000269
RIMXEJYJXDBLIE-UHFFFAOYSA-N
6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene
75906
6-bromohex-1-ene
C=CCCCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.2170g/mL
Boiling Point 47.0°C to 51.0°C (16.0 mmHg)
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)4CH=CH2
Refractive Index 1.4645 to 1.4665 (20°C, 589nm)
Quantity 25 g
Specific Gravity 1.217
Formula Weight 163.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Bromo-1-hexene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 6-Bromo-1-hexene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'