missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-(Boc-amino)quinoline, 97%

Brand:  Alfa Aesar™ H63893.03

Code : NEW

Additional Details : CAS Number : 221070-93-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450849

  • £83.63 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

6-(Boc-amino)quinoline
221070-93-5
244.29
tert-Butyl quinolin-6-ylcarbamate; Quinolin-6-yl-carbamic acid tert-butyl ester

Specifications

6-(Boc-amino)quinoline
Solid
244.29
tert-Butyl quinolin-6-ylcarbamate; Quinolin-6-yl-carbamic acid tert-butyl ester
221070-93-5
97%
1g
Videos
SDS