missing translation for 'onlineSavingsMsg'
Learn More

6-Benzoyl-3-(2-thienyl)-2(1H)-quinoxalinone, 97%, Thermo Scientific™

Catalog Number p-7039316
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1858252-00-2
C19H12N2O2S
332.377
MFCD22988961
DAONFPVIAXDGSI-UHFFFAOYSA-N
6-benzoyl-3-thiophen-2-yl-1h-quinoxalin-2-one, 6-benzoyl-3-2-thienyl-2 1h-quinoxalinone
98000099
6-benzoyl-3-thiophen-2-yl-1H-quinoxalin-2-one
C1=CC=C(C=C1)C(=O)C2=CC3=C(C=C2)NC(=O)C(=N3)C4=CC=CS4
This item is not returnable. View return policy
1858252-00-2
C19H12N2O2S
332.377
MFCD22988961
DAONFPVIAXDGSI-UHFFFAOYSA-N
6-benzoyl-3-thiophen-2-yl-1h-quinoxalin-2-one, 6-benzoyl-3-2-thienyl-2 1h-quinoxalinone
98000099
6-benzoyl-3-thiophen-2-yl-1H-quinoxalin-2-one
C1=CC=C(C=C1)C(=O)C2=CC3=C(C=C2)NC(=O)C(=N3)C4=CC=CS4

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 332.37
Percent Purity 97%
Chemical Name or Material 6-Benzoyl-3-(2-thienyl)-2(1H)-quinoxalinone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'