missing translation for 'onlineSavingsMsg'
Learn More

6-Azaindole, ≥98%, Thermo Scientific Chemicals

Catalog Number p-7055790
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
271-29-4
C7H6N2
118.14
MFCD01686899
XLKDJOPOOHHZAN-UHFFFAOYSA-N
6-azaindole, 1h-pyrrolo 2,3-c pyridine, harmyrine, 1,6-diazaindene, 6h-pyrrolo 2,3-c pyridine, 6-azaindol, pyrrolo 2,3-c pyridine, 6-aza-6h-indole, zlchem 439
9219
1H-pyrrolo[2,3-c]pyridine
N1C=CC2=CC=NC=C12
This item is not returnable. View return policy
271-29-4
C7H6N2
118.14
MFCD01686899
XLKDJOPOOHHZAN-UHFFFAOYSA-N
6-azaindole, 1h-pyrrolo 2,3-c pyridine, harmyrine, 1,6-diazaindene, 6h-pyrrolo 2,3-c pyridine, 6-azaindol, pyrrolo 2,3-c pyridine, 6-aza-6h-indole, zlchem 439
9219
1H-pyrrolo[2,3-c]pyridine
N1C=CC2=CC=NC=C12

6-Azaindole is an indole derivative and useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Azaindole is an indole derivative and useful synthetic intermediate.

Solubility
Soluble in water (hardly), methanol and chloroform.

Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 109685
Sensitivity Air and light sensitive
Solubility Information Soluble in water (hardly),methanol and chloroform.
Formula Weight 118.14
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 6-Azaindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'