missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Azaindole, ≥98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
6-Azaindole is an indole derivative and useful synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Azaindole is an indole derivative and useful synthetic intermediate.
Solubility
Soluble in water (hardly), methanol and chloroform.
Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air.
6-Azaindole is an indole derivative and useful synthetic intermediate.
Solubility
Soluble in water (hardly), methanol and chloroform.
Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air.
Specifications
Quantity | 250 mg |
Beilstein | 109685 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in water (hardly),methanol and chloroform. |
Formula Weight | 118.14 |
Percent Purity | ≥98% |
Physical Form | Solid |
Chemical Name or Material | 6-Azaindole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'