missing translation for 'onlineSavingsMsg'
Learn More

6-Aminopenicillanic acid, 96%, Thermo Scientific Chemicals

Catalog Number 10568863
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
551-16-6
C8H12N2O3S
216.26
MFCD00005176
NGHVIOIJCVXTGV-ALEPSDHESA-N
6-aminopenicillanic acid, 6-apa, penicin, penin, aminopenicillanic acid, 6-aps, phenacyl 6-aminopenicillinate, 6-aminopenicillamine acid, 6-aminopenicillanate, 2s,5r,6r-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid
11082
CHEBI:16705
(2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O
This item is not returnable. View return policy
551-16-6
C8H12N2O3S
216.26
MFCD00005176
NGHVIOIJCVXTGV-ALEPSDHESA-N
6-aminopenicillanic acid, 6-apa, penicin, penin, aminopenicillanic acid, 6-aps, phenacyl 6-aminopenicillinate, 6-aminopenicillamine acid, 6-aminopenicillanate, 2s,5r,6r-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid
11082
CHEBI:16705
(2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 197°C to 203°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 454
Specific Rotation Condition +276.30° (22°C c=1.2,0.1N HCl)
Specific Rotation + 276.3
Formula Weight 216.25
Percent Purity 96%
Physical Form Fine Powder
Chemical Name or Material 6-Aminopenicillanic acid, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Aminopenicillanic acid
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'