missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinic acid methyleester, 97%, ACROS Organics™

Brand:  Acros Organics 366810250

Code : 29

Additional Details : CAS Number : 36052-24-1 Weight : 0.07500kg


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10475745

  • £314.50 / 25g


Estimated Shipment
19-07-2018
Add to basket


Specifications

Chemical Identifiers

6-Aminonicotinic acid methyleester
36052-24-1
100.0
97%
152.15
Methyl 6-aminonicotinate
97%
96.0
≥96.0%
C7H8N2O2
MFCD00797844

Specifications

6-Aminonicotinic acid methyleester
36052-24-1
100.0
Glass bottle
White to light yellow-beige or pink
≥96.0%
25g
152.15
Methyl 6-aminonicotinate
97%
96.0
Authentic
155° to 160°C
Powder
97%
C7H8N2O2
MFCD00797844
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Aminonicotinic acid methyl ester

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS