missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinic acid methyl ester, 97%, Thermo Scientific Chemicals

Catalog Number p-2690880
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
36052-24-1
C7H8N2O2
152.15
MFCD00797844
JACPDLJUQLKABC-UHFFFAOYSA-N
methyl 6-aminonicotinate, 6-aminonicotinic acid methyl ester, 6-amino-nicotinic acid methyl ester, methyl6-aminonicotinate, 2-amino-5-methoxycarbonyl pyridine, methyl 2-amino-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-amino-, methyl ester, methyl 2-aminopyridine-5-carboxylate, 6-aminopyridine-3-carboxylic acid methyl ester
737487
methyl 6-aminopyridine-3-carboxylate
COC(=O)C1=CN=C(C=C1)N
This item is not returnable. View return policy
36052-24-1
C7H8N2O2
152.15
MFCD00797844
JACPDLJUQLKABC-UHFFFAOYSA-N
methyl 6-aminonicotinate, 6-aminonicotinic acid methyl ester, 6-amino-nicotinic acid methyl ester, methyl6-aminonicotinate, 2-amino-5-methoxycarbonyl pyridine, methyl 2-amino-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-amino-, methyl ester, methyl 2-aminopyridine-5-carboxylate, 6-aminopyridine-3-carboxylic acid methyl ester
737487
methyl 6-aminopyridine-3-carboxylate
COC(=O)C1=CN=C(C=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 159°C to 160°C
Color Beige-Yellow to White or Pink
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 152.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Aminonicotinic acid methyl ester, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Aminonicotinic acid methyl ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'