missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinic acid, 98%, ACROS Organics™

Brand:  Acros Organics 202540250

Code : 29

Additional Details : CAS Number : 3167-49-5 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10523761

  • £224.20 / 25g


Estimated Shipment
13-07-2018
Add to basket

Specifications

Chemical Identifiers

6-Aminonicotinic acid
3167-49-5
100.0
98%
H2N(C5H3N)COOH
MFCD00006326
98%
97.5
≥97.5%
C6H6N2O2
138.13
6-Aminopyridine-3-carboxylic acid

Specifications

6-Aminonicotinic acid
3167-49-5
100.0
Glass bottle
Powder
98%
C6H6N2O2
138.13
22, 542
6-Aminopyridine-3-carboxylic acid
98%
97.5
Authentic
White to beige
≥97.5%
25g
H2N(C5H3N)COOH
MFCD00006326
15, 452
Videos
SDS