missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Aminonicotinamide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 245°C to 248°C |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Linear Formula | H2N(C5H3N)CONH2 |
Quantity | 1 g |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 137.4 |
Percent Purity | 98% |
Chemical Name or Material | 6-Aminonicotinamide |
Safety and Handling
Product Identifier
- 6-Aminonicotinamide
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'