missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoindole, 97%, ACROS Organics™

Brand:  Acros Organics 433330050

Code : 29

Additional Details : CAS Number : 5318-27-4 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10482950

  • £164.30 / 5g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

6-Aminoindole
96.0
97%
C8H8N2
MFCD02093959
5318-27-4
100.0
97%
132.17

Specifications

6-Aminoindole
96.0
Authentic
Crystalline Powder
97%
66.0° to 68.0°C
132.17
White to brown
5318-27-4
100.0
Glass bottle
97%
C8H8N2
161.0° to 166.0°C (2.3 Torr)
MFCD02093959
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Aminoindole, 97%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.

SDS