missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoindan-1-one, 97%, ACROS Organics™

Brand:  Acros Organics 432830010

£39.96 valid until 30/06/2017
Log in to get the promotional price



UNSPSC : 12161500

Code : 29

Additional Details : CAS Number : 69975-65-1 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10556830

  • £99.90 / 1g


EU Stock 3
Add to basket


Specifications Description & Specifications

Chemical Identifiers

6-Aminoindan-1-one
96.0
≥96.0% (HPLC)
C9H9NO
MFCD00100747
69975-65-1
100.0
97%
147.18

Specifications

6-Aminoindan-1-one
96.0
Authentic
Solution
97%
155° to 159°C
MFCD00100747
1g
69975-65-1
100.0
Glass bottle
≥96.0% (HPLC)
C9H9NO
147.18
Light brown to black

Safety and Handling

Product Identifier
  • 6-Aminoindan-1-one

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents

Special Offers

Special Offers