missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Aminobenzoxazole, 95%

Brand:  Alfa Aesar™ H34485.06

Code : NEW

Additional Details : CAS Number : 177492-52-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15477437

  • £395.40 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Description

Description

Applications
6-Aminobenzoxazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

6-Aminobenzoxazole
134.1
Benzoxazol-6-amine
177492-52-3
MFCD03791187

Specifications

6-Aminobenzoxazole
Solid
95° to 97°C
134.1
MFCD03791187
Benzoxazol-6-amine
177492-52-3
95%
Orange to brown
5g
Air Sensitive
Videos
SDS