missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Aminobenzo[b]thiophene-1,1-dioxide, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 181.21 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 6-Aminobenzo[b]thiophene 1, 1-dioxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'