missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Amino-7-deazapurine hydrogen sulfate, 97%

Brand:  Alfa Aesar™ H64602.03

Code : NEW

Additional Details : CAS Number : 769951-32-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15431879

  • £23.30 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

6-Amino-7-deazapurine hydrogen sulfate
232.21
4-Amino-7H-pyrrolo[2,3-d]pyrimidine hydrogen sulfate
769951-32-8
MFCD09787504

Specifications

6-Amino-7-deazapurine hydrogen sulfate
Brown
97%
232.21
4-Amino-7H-pyrrolo[2,3-d]pyrimidine hydrogen sulfate
769951-32-8
Powder
1g
MFCD09787504
Videos
SDS