missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-7-deazapurine hydrogen sulfate, 97%, Thermo Scientific™

Catalog Number p-7055895
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
769951-32-8
C6H8N4O4S
232.214
MFCD09787504
ZVFLCRGXDSPVKK-UHFFFAOYSA-N
4-amino-7h-pyrrolo 2,3-d pyrimidine hydrogen sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulphate, 4-amino-7h-pyrrolo 2,3-d pyrimidine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine; sulfuric acid, 7-deazaadenine; sulfuric acid, 1h-pyrrolo 2,3-d pyrimidin-4-amine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulfuric acid, 4-amino-7h-pyrrolo 2,3-d pyrimidinehydrogensulfate, ksc376q6n
11470294
7H-pyrrolo[2,3-d]pyrimidin-4-amine;sulfuric acid
C1=CNC2=C1C(=NC=N2)N.OS(=O)(=O)O
This item is not returnable. View return policy
769951-32-8
C6H8N4O4S
232.214
MFCD09787504
ZVFLCRGXDSPVKK-UHFFFAOYSA-N
4-amino-7h-pyrrolo 2,3-d pyrimidine hydrogen sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulphate, 4-amino-7h-pyrrolo 2,3-d pyrimidine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine; sulfuric acid, 7-deazaadenine; sulfuric acid, 1h-pyrrolo 2,3-d pyrimidin-4-amine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulfuric acid, 4-amino-7h-pyrrolo 2,3-d pyrimidinehydrogensulfate, ksc376q6n
11470294
7H-pyrrolo[2,3-d]pyrimidin-4-amine;sulfuric acid
C1=CNC2=C1C(=NC=N2)N.OS(=O)(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Brown
Quantity 250 mg
Formula Weight 232.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Amino-7-deazapurine hydrogen sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'