missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-5-bromoquinoline, 97%, Thermo Scientific Chemicals

Catalog Number 15419688
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
50358-42-4
C9H7BrN2
223.073
MFCD00234503
MODLGTLYXJGDCH-UHFFFAOYSA-N
12741232
5-bromoquinolin-6-amine
C1=CC2=C(C=CC(=C2Br)N)N=C1
This item is not returnable. View return policy
50358-42-4
C9H7BrN2
223.073
MFCD00234503
MODLGTLYXJGDCH-UHFFFAOYSA-N
12741232
5-bromoquinolin-6-amine
C1=CC2=C(C=CC(=C2Br)N)N=C1

6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

Solubility
Slightly soluble in water. Soluble in alcohol, ether, and carbon disulfide.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Quantity 250 mg
Synonym 6-amino-5-bromoquinoline, 6-quinolinamine, 5-bromo, 5-bromo-6-quinolinamine, 6-quinolinamine,5-bromo, 5-bromo-quinolin-6-ylamine, 6-quinolinamine, 5-bromo-9ci
Solubility Information Slightly soluble in water. Soluble in alcohol,ether,and carbon disulfide.
Formula Weight 223.07
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Amino-5-bromoquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'