missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Amino-5-bromoquinoline, 97%

Brand:  Alfa Aesar™ H63150.06

Code : NEW

Additional Details : CAS Number : 50358-42-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15409688

  • £649.80 / 5g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Description

Description

Applications
6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

Solubility
Slightly soluble in water. Soluble in alcohol, ether, and carbon disulfide.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

6-Amino-5-bromoquinoline
223.07
5-Bromo-quinolin-6-ylamine
50358-42-4
MFCD00234503

Specifications

6-Amino-5-bromoquinoline
Solid
Yellow to brown
5g
5-Bromo-quinolin-6-ylamine
50358-42-4
97%
223.07
MFCD00234503
Videos
SDS