Learn More
Thermo Scientific Maybridge 6-Amino-2-methylnicotinonitrile, 25g, 97%
Brand: Thermo Scientific Maybridge MO01041EB
6-Amino-2-methylnicotinonitrile, 25g, 97%
Code : 31
Additional Details : CAS Number : 183428-90-2 Weight : 0.07500kg
Brand: Thermo Scientific Maybridge MO01041EB
6-Amino-2-methylnicotinonitrile, 25g, 97%
Code : 31
Additional Details : CAS Number : 183428-90-2 Weight : 0.07500kg