missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Amino-2,3-difluorophenol, 95%

Brand:  Alfa Aesar™ H63249.03

Code : NEW

Additional Details : CAS Number : 115551-33-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419848

  • £36.36 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

6-Amino-2,3-difluorophenol
145.11
3,4-Difluoro-2-hydroxyaniline
115551-33-2
MFCD03094310

Specifications

6-Amino-2,3-difluorophenol
Solid
1g
MFCD03094310
115551-33-2
95%
145.11
3,4-Difluoro-2-hydroxyaniline
Videos
SDS