missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-methyl-1H-indazole, 97%, ACROS Organics™

Brand:  Acros Organics 429590010

Code : 29

Additional Details : CAS Number : 74728-65-7 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10106287

  • £64.10 / 1g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

6-Amino-1-methyl-1H-indazole
96.0
97%
147.18
74728-65-7
100.0
C8H9N3
MFCD00456076

Specifications

6-Amino-1-methyl-1H-indazole
96.0
Glass bottle
97%
97%
147.18
74728-65-7
100.0
174.5° to 176.0°C
1g
C8H9N3
MFCD00456076
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Amino-1-methyl-1H-indazole

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS