missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6,7-Dimethoxy-3,4-dihydroisoquinoline, 97%

POA - £188.00

Chemical Identifiers

CAS 3382-18-1
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00266748
InChI Key NSLJVQUDZCZJLK-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-3,4-dihydro-isoquinoline, 3,4-dihydro-6,7-dimethoxyisoquinoline, isoquinoline, 3,4-dihydro-6,7-dimethoxy, pubchem12828, ksc571e8h, 3,4-dihydro-6,7-dimethoxy-isoquinoline, isoquinoline,3,4-dihydro-6,7-dimethoxy, isoquinoline, 3,4-dihydro-6,7-dimethoxy-, hydrochloride 1:1
PubChem CID 30058
IUPAC Name 6,7-dimethoxy-3,4-dihydroisoquinoline
SMILES COC1=CC2=C(C=NCC2)C=C1OC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15411039
SDS View Documents
Alfa Aesar™
H64018.MD
250mg
POA
250mg
View alternative products
Save Up to 
»
N/A Please to purchase this item. Need a web account? Register with us today!
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
15491029
SDS View Documents
Alfa Aesar™
H64018.03
1g
£52.80
1g
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2021
Please to purchase this item. Need a web account? Register with us today!
15401039
SDS View Documents
Alfa Aesar™
H64018.06
5g
£188.00
5g
View alternative products
Save Up to 
»
Estimated Shipment: 25-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

3382-18-1
191.23
NSLJVQUDZCZJLK-UHFFFAOYSA-N
30058
COC1=CC2=C(C=NCC2)C=C1OC
C11H13NO2
MFCD00266748
6,7-dimethoxy-3,4-dihydro-isoquinoline, 3,4-dihydro-6,7-dimethoxyisoquinoline, isoquinoline, 3,4-dihydro-6,7-dimethoxy, pubchem12828, ksc571e8h, 3,4-dihydro-6,7-dimethoxy-isoquinoline, isoquinoline,3,4-dihydro-6,7-dimethoxy, isoquinoline, 3,4-dihydro-6,7-dimethoxy-, hydrochloride 1:1
6,7-dimethoxy-3,4-dihydroisoquinoline

Specifications

6,7-Dimethoxy-3,4-dihydroisoquinoline
155°C to 161°C (1mmHg)
39°C to 40°C
3382-18-1
MFCD00266748
6,7-dimethoxy-3,4-dihydro-isoquinoline, 3,4-dihydro-6,7-dimethoxyisoquinoline, isoquinoline, 3,4-dihydro-6,7-dimethoxy, pubchem12828, ksc571e8h, 3,4-dihydro-6,7-dimethoxy-isoquinoline, isoquinoline,3,4-dihydro-6,7-dimethoxy, isoquinoline, 3,4-dihydro-6,7-dimethoxy-, hydrochloride 1:1
COC1=CC2=C(C=NCC2)C=C1OC
191.23
191.23
97%
1.593
Yellow
250mg
C11H13NO2
138269
NSLJVQUDZCZJLK-UHFFFAOYSA-N
6,7-dimethoxy-3,4-dihydroisoquinoline
30058
Low Melting Solid
SDS
Product Certifications