missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-1-tetralone, 97%, Thermo Scientific Chemicals

Catalog Number p-4229182
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13575-75-2
C12H14O3
206.24
MFCD00134100
YNNJHKOXXBIJKK-UHFFFAOYSA-N
6,7-dimethoxy-1-tetralone, 6,7-dimethoxy-3,4-dihydronaphthalen-1 2h-one, 6,7-dimethoxytetralin-1-one, 6,7-dimethoxy-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 3,4-dihydro-6,7-dimethoxy, 3,4-dihydro-6,7-dimethoxynaphthalen-1 2h-one, 6,7-dimethoxy-2,3,4-trihydronaphthalen-1-one, 7-dimethoxy, 4-dihydro-6, zlchem 843
266816
6,7-dimethoxy-3,4-dihydro-2H-naphthalen-1-one
COC1=CC2=C(C=C1OC)C(=O)CCC2
This item is not returnable. View return policy
13575-75-2
C12H14O3
206.24
MFCD00134100
YNNJHKOXXBIJKK-UHFFFAOYSA-N
6,7-dimethoxy-1-tetralone, 6,7-dimethoxy-3,4-dihydronaphthalen-1 2h-one, 6,7-dimethoxytetralin-1-one, 6,7-dimethoxy-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 3,4-dihydro-6,7-dimethoxy, 3,4-dihydro-6,7-dimethoxynaphthalen-1 2h-one, 6,7-dimethoxy-2,3,4-trihydronaphthalen-1-one, 7-dimethoxy, 4-dihydro-6, zlchem 843
266816
6,7-dimethoxy-3,4-dihydro-2H-naphthalen-1-one
COC1=CC2=C(C=C1OC)C(=O)CCC2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 95.0°C to 98.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 206.24
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 6, 7-Dimethoxy-1-tetralone
Show More Show Less
compliance-icons
Product Identifier
  • 6,7-Dimethoxy-1-tetralone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'