missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one, 98%

Brand:  Alfa Aesar™ H35321.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409005

  • £334.10 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Description

Description

Applications
6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one
172.18
MFCD22988925

Specifications

6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one
Solid
Green
MFCD22988925
1g
98%
172.18
Videos
SDS