missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(2,2,2-Trifluoroethoxy)nicotinic acid, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 221.13 |
Percent Purity | 95% |
Chemical Name or Material | 6-(2, 2, 2-Trifluoroethoxy)nicotinic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'