missing translation for 'onlineSavingsMsg'
Learn More

6,13-Pentacenequinone, 99%, Thermo Scientific Chemicals

Catalog Number p-2690867
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3029-32-1
C22H12O2
308.34
MFCD00003709
UFCVADNIXDUEFZ-UHFFFAOYSA-N
6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa
76415
pentacene-6,13-dione
C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3029-32-1
C22H12O2
308.34
MFCD00003709
UFCVADNIXDUEFZ-UHFFFAOYSA-N
6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa
76415
pentacene-6,13-dione
C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 394.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 07,I,451
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,benzene,pyridine,dioxane,,dimethylformamide,nitrobenzene
Formula Weight 308.34
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 6, 13-Pentacenequinone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 6,13-Pentacenequinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'