missing translation for 'onlineSavingsMsg'
Learn More

6-(1,4-Diazepan-1yl)nicotinonitrile, ≥97%, Thermo Scientific™

Catalog Number p-3075810
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
683274-59-1
C11H14N4
202.261
MFCD01567269
AWTOGESBXYCVMV-UHFFFAOYSA-N
6-1,4-diazepan-1-yl nicotinonitrile, 6-1,4-diazepan-1-yl pyridine-3-carbonitrile, 3-pyridinecarbonitrile,6-hexahydro-1h-1,4-diazepin-1-yl, maybridge4_004605, 6-1,4-diazepan-1yl nicotinonitrile, 6-homopiperazin-1-yl nicotinonitrile, 6-1,4-diazepan-1-yl-3-pyridinecarbonitrile, 6-1,4-diazaperhydroepinyl pyridine-3-carbonitrile
2747101
6-(1,4-diazepan-1-yl)pyridine-3-carbonitrile
C1CNCCN(C1)C2=NC=C(C=C2)C#N
This item is not returnable. View return policy
683274-59-1
C11H14N4
202.261
MFCD01567269
AWTOGESBXYCVMV-UHFFFAOYSA-N
6-1,4-diazepan-1-yl nicotinonitrile, 6-1,4-diazepan-1-yl pyridine-3-carbonitrile, 3-pyridinecarbonitrile,6-hexahydro-1h-1,4-diazepin-1-yl, maybridge4_004605, 6-1,4-diazepan-1yl nicotinonitrile, 6-homopiperazin-1-yl nicotinonitrile, 6-1,4-diazepan-1-yl-3-pyridinecarbonitrile, 6-1,4-diazaperhydroepinyl pyridine-3-carbonitrile
2747101
6-(1,4-diazepan-1-yl)pyridine-3-carbonitrile
C1CNCCN(C1)C2=NC=C(C=C2)C#N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material 6-(1, 4-Diazepan-1-yl)nicotinonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'