missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (5S,6R)-(+)-4-Boc-5,6-diphenyl-2-morpholinone, 99%

Brand:  Alfa Aesar™ H60708.03

Code : NEW

Additional Details : CAS Number : 112741-50-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407198

  • £109.71 / 1g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

(5S,6R)-(+)-4-Boc-5,6-diphenyl-2-morpholinone
353.41
tert-Butyl (2S,3R)-(+)-6-oxo-2,3-diphenylmorpholine-4-carboxylate; (2S,3R)-(+)-4-Boc-6-oxo-2,3-diphenylmorpholine
112741-50-1
MFCD00074953

Specifications

(5S,6R)-(+)-4-Boc-5,6-diphenyl-2-morpholinone
112741-50-1
99%
MFCD00074953
206°C (dec.)
1g
Solid
353.41
tert-Butyl (2S,3R)-(+)-6-oxo-2,3-diphenylmorpholine-4-carboxylate; (2S,3R)-(+)-4-Boc-6-oxo-2,3-diphenylmorpholine
Videos
SDS