missing translation for 'onlineSavingsMsg'
Learn More

5H-Chromeno[2,3-b]pyridin-5-one, 97%, Thermo Scientific™

Catalog Number p-2702843
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6537-46-8
C12H7NO2
MFCD00052253
This item is not returnable. View return policy
6537-46-8
C12H7NO2
MFCD00052253
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 197.19
Percent Purity 97%
Chemical Name or Material 5H-Chromeno[2, 3-b]pyridin-5-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'