missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 5810 Centrifuges with S-4-104 Model Rotor

Use this centrifuge for medium- to high-throughput applications. Eppendorf™ 5810 Centrifuges combine extraordinary versatility and capacity for both tubes and plates with an extraordinary compact footprint.

Brand:  Eppendorf™ 5810000670

Code : NEW

Additional Details : Weight : 89,300.00000kg

 View more versions of this product

Product Code. 15204536

  • £7013.42 / Each


Estimated Shipment
07-02-2018
Add to basket

Specifications

Specifications

68kg
2 to 40°C
230V 50/60Hz
Centrifuge
345mm
535mm
S-4-104 Model Rotor, Adapters for 15mL/50mL Conical Tubes
200 to 14,000rpm
1 to 99min.
4 x 750mL
5810
20,913 x G
<70dB(A)
900W
608mm
Videos