missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 5804R Centrifuges without Rotor

High speed centrifuge for medium capacity needs.

Brand:  Eppendorf™ 5805000017

UNSPSC : 41103904

Code : Z1

Additional Details : Weight : 95.00000kg

 View more versions of this product

Product Code. 10334751

  • £4996.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

4 x 250mL
Yes
230V
80Kg
1 to 99min.
1650W
5804R
550mm
1Each
15 to 35°C
20,913 x G
50 to 60Hz
342mm
<58dB(A)
634mm
230V, 50 to 60Hz, 1650w
80Kg
200 to 14,000rpm
Centrifuge