missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 5424R Centrifuges without Rotor

Equipped for all modern molecular biology applications using Eppendorf and PCR tubes. Eppendorf™ 5424 R Centrifuges are designed for separating liquid substance mixtures with different densities, and analyzing samples from the human body in in-vitro diagnostic applications.

Brand:  Eppendorf™ 5404000219

Code : NEW

Additional Details : Weight : 21.60000kg

 View more versions of this product

Product Code. 10761997

  • £3888.75 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Features state of the art cooling technology to ensure energy efficiency and high temperature accuracy for maximum sample protection
  • OptiBowl™ design for quiet operation, even without a rotor lid
  • Soft-touch one-finger lid closure for ergonomic operation
  • SOFT brake to protect sensitive samples
  • Full digital display with time and speed display
  • Separate short-spin key for quick centrifugation
  • Certified aerosol-tight rotor comes standard for safe centrifugation of hazardous samples
  • Available as a rotary knob version for fast parameter selection, or keypad version for easy cleaning
  • Temperature range from -10  to +40°C
  • Guaranteed temperature of 4°C at maximum rotational speed
  • FastTemp function for quick pre-cooling of the centrifuge, e.g. from ~ 21 to 4°C in only 8 minutes
  • ECO power shut-off engages after 8 hours of non-use to reduce energy consumption (40% over night) and extend compressor life (can be deactivated)
  • The patented compressor technology reduces vibrations and protects your samples
  • Built-in condensation drain to eliminate water accumulation in the rotor chamber
Specifications

Specifications

46.3 lb. (21kg)
230V 50/60Hz
Centrifuge
<54dB(A)
Keypad and GB Plug
350W
Yes
24 x 2mL
5424R
21,130 x G
–11 to 40°C
480mm
100 to 15000rpm
30Sec to 9:59Hr
Videos