Learn More
Cellpath Stainless-steel Reusable Base Molds
Ideal for prostrate and breast cores and can be used with both grid and slatted processing cassettes for both horizontal and vertical sectioning. Cellpath Stainless-steel Reusable Base Moulds are manufactured from high quality stainless steel that put an end to extensive trimming of wax.
Brand: Cellpath GBB-1014-05A
Additional Details : Weight : 1.00000kg
Description
- Made of reusable Stainless steel
- Stackable to reduce storage area
- Internal rounded corners enable easy removal of the wax block
- Features extended edges prevent tipping and provide grip area for ease of handling
- Available in wide range of sizes
- GBB-0714-05A suitable for use with needle and punch biopsies
Specifications
Stainless Steel | |
10 x 24 x 5 mm | |
5/Pk. |