missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ ECL™ start Western Blotting Detection Reagent
Reagent is luminol-based chemiluminescent substrate designed for the detection of proteins immobilized to a membrane
£116.31 - £187.00
Specifications
Product Type | ECL Western Blotting Detection Reagent |
---|---|
For Use With (Application) | Routine and confirmatory Western blotting |
Storage Requirements | 18°C to 30°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15387655
|
Cytiva
RPN3243 |
200 mL |
£116.31
200mL |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
15397655
|
Cytiva
RPN3244 |
400 mL |
£187.00
400mL |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
Amersham™ ECL™ start is a chemiluminescent detection reagent in the Amersham ECL product family that is recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins. Amersham ECL start has a long signal duration, which enables multiple exposures of the membrane plus a convenient time window between experiment and analysis.
- Long signal duration provides assay design flexibility and robust detection of large experimental sets
- Produces low background, regardless of method of detection
- Working solution is stable for five days
- Long shelf life of 18 hours is convenient for those who run Western blots infrequently
- Detection on X-ray film or CCD Imager
- Recommended Membrane: PVDF or nitrocellulose
- For detection of high abundant proteins
- Colorless detection reagent 1 and 2
Applications:
Routine and confirmatory Western blotting
Specifications
ECL Western Blotting Detection Reagent | |
Routine and confirmatory Western blotting | |
18°C to 30°C |